Order Entry
United Kingdom
ContactUsLinkComponent
INCLUDES THE BELOW: CFU 4020 – Walkin Unit Estimated overall dimensions - 4100x2600x2650mm (WxDxH) Estimated useful dimensions - 4000x2000x1950mm (WxDxH) COOLING COIL 1 LED LIGHTING 1 DOUBLE H13 BAG IN/BAG OUT PREFILTERS 1 HEPA FILTERS N.01 LEVEL OF 1 * 1 items
Catalog # FASTFAS/1040R2-CFU
Supplier:  FASTER
New Product
INCLUDES THE BELOW: CFU 4020 – Walkin Unit Estimated overall dimensions - 4100x2600x2650mm (WxDxH) Estimated useful dimensions - 4000x2000x1950mm (WxDxH) COOLING COIL 1 LED LIGHTING 1 DOUBLE H13 BAG IN/BAG OUT PREFILTERS 1 HEPA FILTERS N.01 LEVEL OF 1 * 1 items
Catalog # FASTFAS/1040R2-CFU
Supplier:  FASTER