Order Entry
United Kingdom
ContactUsLinkComponent
Carotegrast methyl (AJM300) is an orally active and selective α4 integrin antagonist. HCA2969, an active metabolite of Carotegrast m ethyl, is a specific and dual α4β1/α4β7 integrin antagonist. Carotegrast methyl prevents the development of colitis i 1 * 10 mg
Catalog # MDEXHY-124290
Supplier:  MEDCHEMEXPRESS
Carotegrast methyl (AJM300) is an orally active and selective α4 integrin antagonist. HCA2969, an active metabolite of Carotegrast m ethyl, is a specific and dual α4β1/α4β7 integrin antagonist. Carotegrast methyl prevents the development of colitis i 1 * 10 mg
Catalog # MDEXHY-124290
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together