Order Entry
United Kingdom
ContactUsLinkComponent
Mevastatin (Compactin) is a first HMG-CoA reductase inhibitor that belongs to the statins class. Mevastatin is a lipid-lowering agen t, and induces apoptosis, arrests cancer cells in G0/G1 phase. Mevastatin also increases endothelial nitric oxide syn 1 * 5 mg
Catalog # MDEXHY-17408
Supplier:  MEDCHEMEXPRESS
Mevastatin (Compactin) is a first HMG-CoA reductase inhibitor that belongs to the statins class. Mevastatin is a lipid-lowering agen t, and induces apoptosis, arrests cancer cells in G0/G1 phase. Mevastatin also increases endothelial nitric oxide syn 1 * 5 mg
Catalog # MDEXHY-17408
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together