Order Entry
United Kingdom
ContactUsLinkComponent
SBI-797812 is an orally active nicotinamide phosphoribosyltransferase (NAMPT) activator. SBI-797812 shifts NAMPT to NMN formation, i ncreases NAMPT affinity for ATP, stabilizes phosphorylated NAMPT, promotes consumption of the pyrophosphate by-produc 1 * 5 mg
Catalog # MDEXHY-126255
Supplier:  MEDCHEMEXPRESS
SBI-797812 is an orally active nicotinamide phosphoribosyltransferase (NAMPT) activator. SBI-797812 shifts NAMPT to NMN formation, i ncreases NAMPT affinity for ATP, stabilizes phosphorylated NAMPT, promotes consumption of the pyrophosphate by-produc 1 * 5 mg
Catalog # MDEXHY-126255
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together