Order Entry
United Kingdom
ContactUsLinkComponent
ALC-0315 is an ionisable aminolipid that is responsible for mRNA compaction and aids mRNA cellular delivery and its cytoplasmic rele ase through suspected endosomal destabilization. ALC-0315 can be used to form lipid nanoparticle (LNP) delivery vehic 1 * 500 mg
Catalog # MDEXHY-138170
Supplier:  MEDCHEMEXPRESS
ALC-0315 is an ionisable aminolipid that is responsible for mRNA compaction and aids mRNA cellular delivery and its cytoplasmic rele ase through suspected endosomal destabilization. ALC-0315 can be used to form lipid nanoparticle (LNP) delivery vehic 1 * 500 mg
Catalog # MDEXHY-138170
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together