Order Entry
United Kingdom
ContactUsLinkComponent
Trimethoprim is a bacteriostatic antibiotic and an orally active dihydrofolate reductase inhibitor. Trimethoprim is active against a wide range of Gram-positive and Gram-negative aerobic bacteria. Trimethoprim has the potential for the research of u 1 * 500 mg
Catalog # MDEXHY-B0510
Supplier:  MEDCHEMEXPRESS
Trimethoprim is a bacteriostatic antibiotic and an orally active dihydrofolate reductase inhibitor. Trimethoprim is active against a wide range of Gram-positive and Gram-negative aerobic bacteria. Trimethoprim has the potential for the research of u 1 * 500 mg
Catalog # MDEXHY-B0510
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together