Order Entry
United Kingdom
ContactUsLinkComponent
Fosfomycin (MK-0955) sodium is a blood-brain barrier penetrating, broad-spectrum antibiotic by irreversibly inhibiting an early stag e in cell wall synthesis. Fosfomycin sodium shows both in vivo and in vitro activity against a wide range of bacteria 1 * 500 mg
  MDEXHY-W016420
Fosfomycin (MK-0955) sodium is a blood-brain barrier penetrating, broad-spectrum antibiotic by irreversibly inhibiting an early stag e in cell wall synthesis. Fosfomycin sodium shows both in vivo and in vitro activity against a wide range of bacteria 1 * 500 mg
  MDEXHY-W016420
Frequently Bought Together