Order Entry
United Kingdom
ContactUsLinkComponent
BDP FL azide is a BDP dye connector containing an azide group capable of Click Chemistry. The green fluorophore is representative of the borodipyrromethane class of fluorescent dyes and has a high quantum yield in aqueous environments, high stabilit 1 * 5 mg
Catalog # MDEXHY-114353
Supplier:  MEDCHEMEXPRESS
BDP FL azide is a BDP dye connector containing an azide group capable of Click Chemistry. The green fluorophore is representative of the borodipyrromethane class of fluorescent dyes and has a high quantum yield in aqueous environments, high stabilit 1 * 5 mg
Catalog # MDEXHY-114353
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together