Order Entry
United Kingdom
ContactUsLinkComponent
Azido-PEG4-Val-Cit-PAB-OH is a cleavable 4 unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs)[1]. Azido-PE G4-Val-Cit-PAB-OH is also a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[2]. Azido-PEG4-Val-C 1 * 50 mg
Catalog # MDEXHY-140149
Supplier:  MEDCHEMEXPRESS
Azido-PEG4-Val-Cit-PAB-OH is a cleavable 4 unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs)[1]. Azido-PE G4-Val-Cit-PAB-OH is also a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[2]. Azido-PEG4-Val-C 1 * 50 mg
Catalog # MDEXHY-140149
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together