Order Entry
United Kingdom
ContactUsLinkComponent
PROTAC TYK2 degradation agent1 is a potent and subtype-selective TYK2 degrader. PROTAC TYK2 degradation agent1 has TYK2 degradation activity with DC50 value of 14 nM. PROTAC TYK2 degradation agent1 can be used for the research of autoimmune disease[1 1 * 50 mg
Catalog # MDEXHY-152227
Supplier:  MEDCHEMEXPRESS
PROTAC TYK2 degradation agent1 is a potent and subtype-selective TYK2 degrader. PROTAC TYK2 degradation agent1 has TYK2 degradation activity with DC50 value of 14 nM. PROTAC TYK2 degradation agent1 can be used for the research of autoimmune disease[1 1 * 50 mg
Catalog # MDEXHY-152227
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together