Order Entry
United Kingdom
ContactUsLinkComponent
ODIPY Green 8-P2M is a novel thiol-reactive fluorescence probe based on the BODIPY fluorophore, the fluorescence is strongly quenche d by d-PeT and then can be restored after reaction with thiol, resulting in an extremely high signal-to-noise ratio. 1 * 10 mg
Catalog # MDEXHY-D1590
Supplier:  MEDCHEMEXPRESS
ODIPY Green 8-P2M is a novel thiol-reactive fluorescence probe based on the BODIPY fluorophore, the fluorescence is strongly quenche d by d-PeT and then can be restored after reaction with thiol, resulting in an extremely high signal-to-noise ratio. 1 * 10 mg
Catalog # MDEXHY-D1590
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together