Order Entry
United Kingdom
ContactUsLinkComponent
FT3967385 is a novel USP30 inhibitor that recapitulates genetic loss of USP30 and sets the trigger for PINK1-PARKIN amplification of mitochondrial ubiquitylation. 1 * 100 mg
Catalog # MDEXHY-145337
Supplier:  MEDCHEMEXPRESS
FT3967385 is a novel USP30 inhibitor that recapitulates genetic loss of USP30 and sets the trigger for PINK1-PARKIN amplification of mitochondrial ubiquitylation. 1 * 100 mg
Catalog # MDEXHY-145337
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together