Order Entry
United Kingdom
ContactUsLinkComponent
Bersiporocin is a prolyl-tRNA synthetase inhibitor. Bersiporocin has an IC50 of ≤100 nM for phosphoribosylpyrophosphate synthetase ( PRS). Bersiporocin can be used for the research of antifibrotic. 1 * 1 mg
Catalog # MDEXHY-145555
Supplier:  MEDCHEMEXPRESS
Bersiporocin is a prolyl-tRNA synthetase inhibitor. Bersiporocin has an IC50 of ≤100 nM for phosphoribosylpyrophosphate synthetase ( PRS). Bersiporocin can be used for the research of antifibrotic. 1 * 1 mg
Catalog # MDEXHY-145555
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together