Order Entry
United Kingdom
ContactUsLinkComponent
UCL-TRO-1938 is a potent allosteric activator of PI3Kα with an EC50 value of approximately 60 μM. UCL-TRO-1938 can induce cell proli feration and has cardioprotective and neural regeneration effects[1]. 1 * 25 mg
Catalog # MDEXHY-154848
Supplier:  MEDCHEMEXPRESS
UCL-TRO-1938 is a potent allosteric activator of PI3Kα with an EC50 value of approximately 60 μM. UCL-TRO-1938 can induce cell proli feration and has cardioprotective and neural regeneration effects[1]. 1 * 25 mg
Catalog # MDEXHY-154848
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together