Order Entry
United Kingdom
ContactUsLinkComponent
Pyrimethamine (Pirimecidan) is a potent, orally active dihydrofolate reductase (DHFR) inhibitor. Pyrimethamine is an antimalarial ag ent. Pyrimethamine affects the nucleoprotein metabolism of malarial parasites by interference in the folic–folinic ac 1 * 500 mg
Catalog # MDEXHY-18062
Supplier:  MEDCHEMEXPRESS
Pyrimethamine (Pirimecidan) is a potent, orally active dihydrofolate reductase (DHFR) inhibitor. Pyrimethamine is an antimalarial ag ent. Pyrimethamine affects the nucleoprotein metabolism of malarial parasites by interference in the folic–folinic ac 1 * 500 mg
Catalog # MDEXHY-18062
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together