Order Entry
United Kingdom
ContactUsLinkComponent
Avasimibe (CI-1011 PD-148515) is an orally active acyl coenzyme A-cholesterol acyltransferase (ACAT also called SOAT)) inhibitor w ith IC50s of 24 and 9.2 µM for ACAT1 and ACAT2, respectively. Avasimibe can be used for the research of prostate canc 1 * 50 mg
Catalog # MDEXHY-13215
Supplier:  MEDCHEMEXPRESS
Avasimibe (CI-1011 PD-148515) is an orally active acyl coenzyme A-cholesterol acyltransferase (ACAT also called SOAT)) inhibitor w ith IC50s of 24 and 9.2 µM for ACAT1 and ACAT2, respectively. Avasimibe can be used for the research of prostate canc 1 * 50 mg
Catalog # MDEXHY-13215
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together