Order Entry
United Kingdom
ContactUsLinkComponent
YM-201636 is a potent and selective PIKfyve inhibitor with an IC50 of 33 nM. YM-201636 also inhibits p110α with an IC50 of 3.3 μM. Y M-201636 inhibits retroviral replication. 1 * 25 mg
Catalog # MDEXHY-13228
Supplier:  MEDCHEMEXPRESS
YM-201636 is a potent and selective PIKfyve inhibitor with an IC50 of 33 nM. YM-201636 also inhibits p110α with an IC50 of 3.3 μM. Y M-201636 inhibits retroviral replication. 1 * 25 mg
Catalog # MDEXHY-13228
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together