Order Entry
United Kingdom
ContactUsLinkComponent
Taselisib (GDC-0032) is a potent PI3K inhibitor targets PIK3CA mutations, with Kis of 0.12 nM, 0.29 nM, 0.97 nM, and 9.1 nM for PI3K δ, PI3Kα, PI3Kγ and PI3Kβ, respectively. 1 * 200 mg
Catalog # MDEXHY-13898
Supplier:  MEDCHEMEXPRESS
Taselisib (GDC-0032) is a potent PI3K inhibitor targets PIK3CA mutations, with Kis of 0.12 nM, 0.29 nM, 0.97 nM, and 9.1 nM for PI3K δ, PI3Kα, PI3Kγ and PI3Kβ, respectively. 1 * 200 mg
Catalog # MDEXHY-13898
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together