Order Entry
United Kingdom
ContactUsLinkComponent
Z-IETD-FMK (Z-IE(OMe)TD(OMe)-FMK) is a selective and cell permeable caspase-8 inhibitor[1]. Z-IETD-FMK is also a granzyme B inhibito r[5]. 1 * 5 mg
Catalog # MDEXHY-101297
Supplier:  MEDCHEMEXPRESS
Z-IETD-FMK (Z-IE(OMe)TD(OMe)-FMK) is a selective and cell permeable caspase-8 inhibitor[1]. Z-IETD-FMK is also a granzyme B inhibito r[5]. 1 * 5 mg
Catalog # MDEXHY-101297
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together