Order Entry
United Kingdom
ContactUsLinkComponent
GOT1 inhibitor-1 (compound 2c), a tryptamine-based derivative, acts as a novel, potent and non-covalent inhibitor of glutamate oxalo acetate transaminase 1 (GOT1) with an IC50 of 8.2 uM. GOT1 plays an important role in energy metabolism and Reactive 1 * 50 mg
Catalog # MDEXHY-122723
Supplier:  MEDCHEMEXPRESS
GOT1 inhibitor-1 (compound 2c), a tryptamine-based derivative, acts as a novel, potent and non-covalent inhibitor of glutamate oxalo acetate transaminase 1 (GOT1) with an IC50 of 8.2 uM. GOT1 plays an important role in energy metabolism and Reactive 1 * 50 mg
Catalog # MDEXHY-122723
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together