Order Entry
United Kingdom
ContactUsLinkComponent
4-Azido-L-phenylalanine is an unnatural amino acid, which is used as an effective vibrational reporter of local protein environments . 4-Azido-L-phenylalanine is a click chemistry reagent, it contains an Azide group and can undergo copper-catalyzed a 1 * 100 mg
Catalog # MDEXHY-16714
Supplier:  MEDCHEMEXPRESS
4-Azido-L-phenylalanine is an unnatural amino acid, which is used as an effective vibrational reporter of local protein environments . 4-Azido-L-phenylalanine is a click chemistry reagent, it contains an Azide group and can undergo copper-catalyzed a 1 * 100 mg
Catalog # MDEXHY-16714
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together