Order Entry
United Kingdom
ContactUsLinkComponent
Salcaprozate sodium (SNAC), an oral absorption promoter, and has the potential as a delivery agent for oral forms of heparin and ins ulin. Salcaprozate sodium could increase passive transcellular permeation across small intestinal epithelia based on 1 * 250 mg
Catalog # MDEXHY-114299
Supplier:  MEDCHEMEXPRESS
Salcaprozate sodium (SNAC), an oral absorption promoter, and has the potential as a delivery agent for oral forms of heparin and ins ulin. Salcaprozate sodium could increase passive transcellular permeation across small intestinal epithelia based on 1 * 250 mg
Catalog # MDEXHY-114299
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together