BI-4916 is a proagent of BI-4924. BI-4924 is a NADH/NAD+-competitive PHGDH inhibitor[1]. 1 * 25 mg
MDEXHY-126253
BI-4916 is a proagent of BI-4924. BI-4924 is a NADH/NAD+-competitive PHGDH inhibitor[1]. 1 * 25 mg
MDEXHY-126253
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection