Order Entry
United Kingdom
ContactUsLinkComponent
PND-1186 (VS-4718) is a potent, highly-specific and reversible inhibitor of FAK with an IC50 of 1.5 nM. PND-1186 selectively promote s tumor cell apoptosis[1]. 1 * 10 mg
  MDEXHY-13917
PND-1186 (VS-4718) is a potent, highly-specific and reversible inhibitor of FAK with an IC50 of 1.5 nM. PND-1186 selectively promote s tumor cell apoptosis[1]. 1 * 10 mg
  MDEXHY-13917
Frequently Bought Together