Order Entry
United Kingdom
ContactUsLinkComponent
Danuglipron (PF-06882961) is an orally active glucagon-like peptide-1 receptor (GLP-1R) agonist. Danuglipron has the potential for t ype 2 diabetes research[1][2]. 1 * 5 mg
Catalog # MDEXHY-125824
Supplier:  MEDCHEMEXPRESS
Danuglipron (PF-06882961) is an orally active glucagon-like peptide-1 receptor (GLP-1R) agonist. Danuglipron has the potential for t ype 2 diabetes research[1][2]. 1 * 5 mg
Catalog # MDEXHY-125824
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together