Order Entry
United Kingdom
ContactUsLinkComponent
Cirtuvivint (SM08502) is a potent and orally active CDC-like kinase (CLK) inhibitor. Cirtuvivint can be used for solid tumors resear ch[1]. 1 * 100 mg
Catalog # MDEXHY-137435
Supplier:  MEDCHEMEXPRESS
Cirtuvivint (SM08502) is a potent and orally active CDC-like kinase (CLK) inhibitor. Cirtuvivint can be used for solid tumors resear ch[1]. 1 * 100 mg
Catalog # MDEXHY-137435
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together