Order Entry
United Kingdom
ContactUsLinkComponent
KGF-2/FGF-10 Protein, Human (169a.a) is a heparin-binding protein that binds to FGF2 IIIb and FGFR1III-b receptors, promotes the gro wth, proliferation, and differentiation of epithelial cells and can enhance corneal wound healing. 1 * 500 µG
  MDEXHY-P7048
KGF-2/FGF-10 Protein, Human (169a.a) is a heparin-binding protein that binds to FGF2 IIIb and FGFR1III-b receptors, promotes the gro wth, proliferation, and differentiation of epithelial cells and can enhance corneal wound healing. 1 * 500 µG
  MDEXHY-P7048
Frequently Bought Together