Order Entry
United Kingdom
ContactUsLinkComponent
Rosopatamab (HuJ591) is a humanized antiPSMA IgG1 monoclonal antibody that can be used in cancer research, especially prostate cance r. Rosopatamab can be linked to the low-energy beta-emitting radioisotope lutetium-177 (177Lu) through the bifunction 1 * 5 mg
Catalog # MDEXHY-P99239
Supplier:  MEDCHEMEXPRESS
Rosopatamab (HuJ591) is a humanized antiPSMA IgG1 monoclonal antibody that can be used in cancer research, especially prostate cance r. Rosopatamab can be linked to the low-energy beta-emitting radioisotope lutetium-177 (177Lu) through the bifunction 1 * 5 mg
Catalog # MDEXHY-P99239
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together