Order Entry
United Kingdom
ContactUsLinkComponent
Aminooxyacetic acid (Carboxymethoxylamine) hemihydrochloride is a malate-aspartate shuttle (MAS) inhibitor which also inhibits the G ABA degradating enzyme GABA-T. 1 * 1 g
Catalog # MDEXHY-107994
Supplier:  MEDCHEMEXPRESS
Aminooxyacetic acid (Carboxymethoxylamine) hemihydrochloride is a malate-aspartate shuttle (MAS) inhibitor which also inhibits the G ABA degradating enzyme GABA-T. 1 * 1 g
Catalog # MDEXHY-107994
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together