Order Entry
United Kingdom
ContactUsLinkComponent
Imatinib Mesylate (STI571 Mesylate) is a tyrosine kinases inhibitor that inhibits c-Kit, Bcr-Abl, and PDGFR (IC50=100 nM) tyrosine k inases. 1 * 200 mg
Catalog # MDEXHY-50946
Supplier:  MEDCHEMEXPRESS
Imatinib Mesylate (STI571 Mesylate) is a tyrosine kinases inhibitor that inhibits c-Kit, Bcr-Abl, and PDGFR (IC50=100 nM) tyrosine k inases. 1 * 200 mg
Catalog # MDEXHY-50946
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together