Order Entry
United Kingdom
ContactUsLinkComponent
C646 is a selective and competitive histone acetyltransferase p300 inhibitor with Ki of 400 nM, and is less potent for other acetylt ransferases[1]. 1 * 10 mg
  MDEXHY-13823
C646 is a selective and competitive histone acetyltransferase p300 inhibitor with Ki of 400 nM, and is less potent for other acetylt ransferases[1]. 1 * 10 mg
  MDEXHY-13823
Frequently Bought Together