Order Entry
United Kingdom
ContactUsLinkComponent
TCEP hydrochloride (Tris(2-carboxyethyl)phosphine hydrochloride) is a non-thiol reducing agent that is more stable and produces a fa ster S-S reductive reaction than other chemical reductants. TCEP hydrochloride is a trialkylphosphine, selectively re 1 * 1 g
Catalog # MDEXHY-W011500
Supplier:  MEDCHEMEXPRESS
TCEP hydrochloride (Tris(2-carboxyethyl)phosphine hydrochloride) is a non-thiol reducing agent that is more stable and produces a fa ster S-S reductive reaction than other chemical reductants. TCEP hydrochloride is a trialkylphosphine, selectively re 1 * 1 g
Catalog # MDEXHY-W011500
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together