Order Entry
United Kingdom
ContactUsLinkComponent
SB-590885 is a potent?B-Raf?inhibitor with?Ki?of 0.16 nM, and has 11-fold greater selectivity for B-Raf over c-Raf, without inhibiti on to other human kinases. 1 * 100 mg
Catalog # MDEXHY-10966
Supplier:  MEDCHEMEXPRESS
SB-590885 is a potent?B-Raf?inhibitor with?Ki?of 0.16 nM, and has 11-fold greater selectivity for B-Raf over c-Raf, without inhibiti on to other human kinases. 1 * 100 mg
Catalog # MDEXHY-10966
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together