Order Entry
United Kingdom
ContactUsLinkComponent
Bisindolylmaleimide I (GF109203X) is a cell-permeable and reversible PKC inhibitor (IC50 of 20 nM, 17 nM, 16 nM, and 20 nM for PKCα, PKCβI, PKCβII, and PKCγ. Bisindolylmaleimide I is also a GSK-3 inhibitor[1][2][3]. 1 * 50 mg
Catalog # MDEXHY-13867
Supplier:  MEDCHEMEXPRESS
Bisindolylmaleimide I (GF109203X) is a cell-permeable and reversible PKC inhibitor (IC50 of 20 nM, 17 nM, 16 nM, and 20 nM for PKCα, PKCβI, PKCβII, and PKCγ. Bisindolylmaleimide I is also a GSK-3 inhibitor[1][2][3]. 1 * 50 mg
Catalog # MDEXHY-13867
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together