Order Entry
United Kingdom
ContactUsLinkComponent
PKR-IN-C16 (Compound C16) is a specific double-stranded RNA-dependent protein kinase (PKR) inhibitor. PKR-IN-C16 shows promising neu roprotective properties and can rescue acute brain lesions[1]. 1 * 100 mg
Catalog # MDEXHY-13977A
Supplier:  MEDCHEMEXPRESS
PKR-IN-C16 (Compound C16) is a specific double-stranded RNA-dependent protein kinase (PKR) inhibitor. PKR-IN-C16 shows promising neu roprotective properties and can rescue acute brain lesions[1]. 1 * 100 mg
Catalog # MDEXHY-13977A
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together