Order Entry
United Kingdom
ContactUsLinkComponent
EX229, a Benzimidazole derivative, is a potent and allosteric activator of AMP-activated protein kinase (AMPK), with Kds of 0.06 μM, 0.06 μM and 0.51 μM for α1β1γ1, α2β1γ1 and α1β2γ1 in biolayer interferometry, respectively. 1 * 50 mg
Catalog # MDEXHY-112769
Supplier:  MEDCHEMEXPRESS
EX229, a Benzimidazole derivative, is a potent and allosteric activator of AMP-activated protein kinase (AMPK), with Kds of 0.06 μM, 0.06 μM and 0.51 μM for α1β1γ1, α2β1γ1 and α1β2γ1 in biolayer interferometry, respectively. 1 * 50 mg
Catalog # MDEXHY-112769
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together