Order Entry
United Kingdom
ContactUsLinkComponent
(-)-DHMEQ (Dehydroxymethylepoxyquinomicin) is a potent, selective and irreversible NF-κB inhibitor that covalently binds to a cystei ne residue. (-)-DHMEQ inhibits nuclear translocation of NF-κB and shows anti-inflammatory and anticancer activity[1][ 1 * 10 mg
Catalog # MDEXHY-14645
Supplier:  MEDCHEMEXPRESS
(-)-DHMEQ (Dehydroxymethylepoxyquinomicin) is a potent, selective and irreversible NF-κB inhibitor that covalently binds to a cystei ne residue. (-)-DHMEQ inhibits nuclear translocation of NF-κB and shows anti-inflammatory and anticancer activity[1][ 1 * 10 mg
Catalog # MDEXHY-14645
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together