Order Entry
United Kingdom
ContactUsLinkComponent
Eculizumab (Anti-Human C5, Humanized Antibody) is a long-acting humanized monoclonal antibody targeted against complement C5. Eculiz umab inhibits the cleavage of C5 into C5a and C5b and hence inhibits deployment of the terminal complement system inc 1 * 10 mg
Catalog # MDEXHY-P9914
Supplier:  MEDCHEMEXPRESS
Eculizumab (Anti-Human C5, Humanized Antibody) is a long-acting humanized monoclonal antibody targeted against complement C5. Eculiz umab inhibits the cleavage of C5 into C5a and C5b and hence inhibits deployment of the terminal complement system inc 1 * 10 mg
Catalog # MDEXHY-P9914
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together