Order Entry
United Kingdom
ContactUsLinkComponent
Atirmociclib (PF-07220060 example A94) is a potent CDK4/CDK6 inhibitor with a Ki of 0.6 nM and 13.9 nM for CDK4/Cyclin D1 and CDK6/ Cyclin D3, respectively[1]. 1 * 1 mg
Catalog # MDEXHY-139450
Supplier:  MEDCHEMEXPRESS
Atirmociclib (PF-07220060 example A94) is a potent CDK4/CDK6 inhibitor with a Ki of 0.6 nM and 13.9 nM for CDK4/Cyclin D1 and CDK6/ Cyclin D3, respectively[1]. 1 * 1 mg
Catalog # MDEXHY-139450
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together