Order Entry
United Kingdom
ContactUsLinkComponent
Tomivosertib (eFT508) is a potent, highly selective, and orally active MNK1 and MNK2 inhibitor, with IC50s of 1-2 nM against both is oforms. Tomivosertib (eFT508) treatment leads to a dose-dependent reduction in eIF4E phosphorylation at serine 209 (I 1 * 100 mg
Catalog # MDEXHY-100022
Supplier:  MEDCHEMEXPRESS
Tomivosertib (eFT508) is a potent, highly selective, and orally active MNK1 and MNK2 inhibitor, with IC50s of 1-2 nM against both is oforms. Tomivosertib (eFT508) treatment leads to a dose-dependent reduction in eIF4E phosphorylation at serine 209 (I 1 * 100 mg
Catalog # MDEXHY-100022
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together