Order Entry
United Kingdom
ContactUsLinkComponent
RapaLink-1, the third-generation bivalent mTOR inhibitor, combines Rapamycin (HY-10219) with MLN0128 (HY-13328, a second-generation mTOR kinase inhibitor) by an inert chemical linker. RapaLink-1 shows better efficacy than Rapamycin or mTOR kinase inh 1 * 10 mg
Catalog # MDEXHY-111373
Supplier:  MEDCHEMEXPRESS
RapaLink-1, the third-generation bivalent mTOR inhibitor, combines Rapamycin (HY-10219) with MLN0128 (HY-13328, a second-generation mTOR kinase inhibitor) by an inert chemical linker. RapaLink-1 shows better efficacy than Rapamycin or mTOR kinase inh 1 * 10 mg
Catalog # MDEXHY-111373
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together