Order Entry
United Kingdom
ContactUsLinkComponent
Octreotide acetate, a long-acting synthetic analog of native somatostatin, inhibits growth hormone, glucagon, and insulin more poten tly. 1 * 10 mg
Catalog # MDEXHY-17365
Supplier:  MEDCHEMEXPRESS
Octreotide acetate, a long-acting synthetic analog of native somatostatin, inhibits growth hormone, glucagon, and insulin more poten tly. 1 * 10 mg
Catalog # MDEXHY-17365
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together