Order Entry
United Kingdom
ContactUsLinkComponent
STX-478 (compound 80) is an oral CNS-penetrant allosteric mutant-selective PI3Kα inhibitor. STX-478 shows robust and durable tumor r egression and can be used in cancer research[1]. 1 * 50 mg
Catalog # MDEXHY-156681
Supplier:  MEDCHEMEXPRESS
STX-478 (compound 80) is an oral CNS-penetrant allosteric mutant-selective PI3Kα inhibitor. STX-478 shows robust and durable tumor r egression and can be used in cancer research[1]. 1 * 50 mg
Catalog # MDEXHY-156681
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together