Order Entry
United Kingdom
ContactUsLinkComponent
Trametinib (DMSO solvate) (GSK-1120212 (DMSO solvate) JTP-74057 (DMSO solvate)) is an orally active MEK inhibitor that inhibits MEK1 and MEK2 with IC50s of about 2 nM. Trametinib (DMSO solvate) activates autophagy and induces apoptosis[1][2]. 1 * 5 mg
Catalog # MDEXHY-10999A
Supplier:  MEDCHEMEXPRESS
Trametinib (DMSO solvate) (GSK-1120212 (DMSO solvate) JTP-74057 (DMSO solvate)) is an orally active MEK inhibitor that inhibits MEK1 and MEK2 with IC50s of about 2 nM. Trametinib (DMSO solvate) activates autophagy and induces apoptosis[1][2]. 1 * 5 mg
Catalog # MDEXHY-10999A
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together