Order Entry
United Kingdom
ContactUsLinkComponent
Cladribine (2-Chloro-2′-deoxyadenosine), a purine nucleoside analog, is an orally active adenosine deaminase inhibitor. Cladribine f unctions as an inhibitor of DNA synthesis to block the repair of the damaged DNA. Cladribine can inhibit DNA methylat 1 * 5 mg
Catalog # MDEXHY-13599
Supplier:  MEDCHEMEXPRESS
Cladribine (2-Chloro-2′-deoxyadenosine), a purine nucleoside analog, is an orally active adenosine deaminase inhibitor. Cladribine f unctions as an inhibitor of DNA synthesis to block the repair of the damaged DNA. Cladribine can inhibit DNA methylat 1 * 5 mg
Catalog # MDEXHY-13599
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together