Order Entry
United Kingdom
ContactUsLinkComponent
Ribavirin (ICN-1229) is an antiviral agent against a broad spectrum of viruses including HCV, HIVl, and RSV. Ribavirin also has anti -orthopoxvirus and anti-variola activities. 1 * 500 mg
Catalog # MDEXHY-B0434
Supplier:  MEDCHEMEXPRESS
Ribavirin (ICN-1229) is an antiviral agent against a broad spectrum of viruses including HCV, HIVl, and RSV. Ribavirin also has anti -orthopoxvirus and anti-variola activities. 1 * 500 mg
Catalog # MDEXHY-B0434
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together