Order Entry
United Kingdom
ContactUsLinkComponent
Dantrolene is an orally active, non-competitive glutathione reductase inhibitor with a Ki of 111.6 μM and an IC50 of 52.3 μM. Dantro lene is also a calcium channel protein inhibitor. Dantrolene inhibits the release of Ca2+ from RyR1 and RyR3, which c 1 * 5 mg
Catalog # MDEXHY-12542
Supplier:  MEDCHEMEXPRESS
Dantrolene is an orally active, non-competitive glutathione reductase inhibitor with a Ki of 111.6 μM and an IC50 of 52.3 μM. Dantro lene is also a calcium channel protein inhibitor. Dantrolene inhibits the release of Ca2+ from RyR1 and RyR3, which c 1 * 5 mg
Catalog # MDEXHY-12542
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together