Order Entry
United Kingdom
ContactUsLinkComponent
GSK1016790A is a potent and selective transient receptor potential vanilloid 4 (TRPV4) channel agonist. GSK1016790A can elicit Ca2+ influx and elevate intracellular Ca2+ in HEK cells[1][2]. 1 * 50 mg
Catalog # MDEXHY-19608
Supplier:  MEDCHEMEXPRESS
GSK1016790A is a potent and selective transient receptor potential vanilloid 4 (TRPV4) channel agonist. GSK1016790A can elicit Ca2+ influx and elevate intracellular Ca2+ in HEK cells[1][2]. 1 * 50 mg
Catalog # MDEXHY-19608
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together