Order Entry
United Kingdom
ContactUsLinkComponent
Compound E is a γ-secretase inhibitor. Compound E bloks β-amyloid(40), β-amyloid(42), and Notch γ-secretase cleavage with IC50s of 0 .24, 0.37, 0.32 nM, respectively. 1 * 50 mg
Catalog # MDEXHY-14176
Supplier:  MEDCHEMEXPRESS
Compound E is a γ-secretase inhibitor. Compound E bloks β-amyloid(40), β-amyloid(42), and Notch γ-secretase cleavage with IC50s of 0 .24, 0.37, 0.32 nM, respectively. 1 * 50 mg
Catalog # MDEXHY-14176
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together