Order Entry
United Kingdom
ContactUsLinkComponent
Oritavancin diphosphate (LY333328 diphosphate), a semisynthetic derivative of Vancomycin (HY-B0671), is an orally active glycopeptid e antibiotic with bactericidal activity against gram-positive organisms. Oritavancin diphosphate shows antibacterial 1 * 2 mg
Catalog # MDEXHY-B1831A
Supplier:  MEDCHEMEXPRESS
Oritavancin diphosphate (LY333328 diphosphate), a semisynthetic derivative of Vancomycin (HY-B0671), is an orally active glycopeptid e antibiotic with bactericidal activity against gram-positive organisms. Oritavancin diphosphate shows antibacterial 1 * 2 mg
Catalog # MDEXHY-B1831A
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together